Learn More
Chromium carbide, 99.5% (metals basis), Thermo Scientific Chemicals
£35.60 - £545.00
Chemical Identifiers
CAS | 12105-81-6 |
---|---|
Molecular Formula | Cr23C6 |
MDL Number | MFCD00678358 |
Synonym | bis methane trichromium |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11334368
|
Thermo Scientific Alfa Aesar
042313.09 |
10 g |
£35.60
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11324368
|
Thermo Scientific Alfa Aesar
042313.18 |
50 g |
£134.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11344368
|
Thermo Scientific Alfa Aesar
042313.30 |
250 g |
£545.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chromium carbide is useful in the surface treatment of metal components. Chromium carbide is used to coat the surface of another metal in a technique known as thermal spraying. Chromium carbide is used as an additive in cutting tools made out of cemented carbides, in order to improve toughness by preventing the growth of large grains.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12105-81-6 | |
Cr23C6 | |
MFCD00678358 | |
bis methane trichromium |
Specifications
12105-81-6 | |
Powder | |
Cr23C6 | |
bis methane trichromium | |
1267.97 | |
Gray powder |
Gray | |
99.5% (Metal basis) | |
MFCD00678358 | |
Reacts with water. | |
Odorless | |
Chromium carbide |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: III
missing translation for 'einecsNumber' : 235-160-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only