missing translation for 'onlineSavingsMsg'
Learn More

Chromium hexacarbonyl, 99+%

Product Code. 10542744 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10542744

Brand: Thermo Scientific Acros 221050500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13007-92-6 | C6CrO6 | 220.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13007-92-6
Molecular Formula C6CrO6
Molecular Weight (g/mol) 220.06
MDL Number MFCD00010945
InChI Key KOTQLLUQLXWWDK-UHFFFAOYSA-N
Synonym chromium hexacarbonyl, chromium carbonyl, chromium 0 hexacarbonyl, carbon monoxide; chromium, hexacarbonyl chromium, unii-920f3ejx2p, ccris 3186, chromium carbonyl oc-6-11, hexacarbonylchromium 10g
PubChem CID 25589
ChEBI CHEBI:33031
IUPAC Name carbon monoxide;chromium
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr]

Specifications

Boiling Point 220°C
Melting Point 150°C
Color White to Yellow
Physical Form Crystalline Powder and/or Crystals
Quantity 50 g
Assay Percent Range 99+%
Linear Formula Cr(CO)6
Merck Index 15, 2236
Solubility Information Solubility in water: insoluble. Other solubilities: sparingly soluble in organic solvents
Formula Weight 220.06
Percent Purity ≥99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Chromium hexacarbonyl
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.