Learn More
Chromium(III) chloride hexahydrate, 98%, Thermo Scientific Chemicals
£46.60 - £228.00
Chemical Identifiers
CAS | 10060-12-5 |
---|---|
Molecular Formula | Cl3CrH12O6 |
Molecular Weight (g/mol) | 266.44 |
MDL Number | MFCD00149660 |
InChI Key | LJAOOBNHPFKCDR-UHFFFAOYSA-K |
Synonym | chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech |
PubChem CID | 104957 |
ChEBI | CHEBI:53442 |
IUPAC Name | chromium(3+) hexahydrate trichloride |
SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10286790
|
Thermo Scientific Acros
213365000 |
500 g | Plastic bottle |
£46.60
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10044103
|
Thermo Scientific Acros
213360051 |
5 kg | Plastic drum |
£228.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
10060-12-5 | |
266.44 | |
LJAOOBNHPFKCDR-UHFFFAOYSA-K | |
104957 | |
chromium(3+) hexahydrate trichloride |
Cl3CrH12O6 | |
MFCD00149660 | |
chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech | |
CHEBI:53442 | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3] |
Specifications
83.0°C | |
Green | |
98% | |
[Cr(H2O)4Cl2]Cl·2H2O | |
14, 2222 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, slightly soluble in acetone, practically insoluble in ether | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3] | |
266.44 | |
CHEBI:53442 | |
98% |
10060-12-5 | |
Crystalline Powder | |
Cl3CrH12O6 | |
MFCD00149660 | |
chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech | |
LJAOOBNHPFKCDR-UHFFFAOYSA-K | |
chromium(3+) hexahydrate trichloride | |
104957 | |
266.44 | |
Chromium(III) chloride hexahydrate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
May be corrosive to metals.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Absorb spillage to prevent material damage.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SK
GHS Signal Word: Warning
- Chromium(III) chloride hexahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 C
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Chromium (III) Chloride Hexahydrate, Chromic chloride
RUO – Research Use Only