missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium(III) chloride hexahydrate, 99.5% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Chromium(III) chloride hexahydrate is used as a catalyst, as a mordant in printing and dyeing industry. It is involved in the preparation of anhydrous tetrahydrofuran complex of chromium(III) chloride by reacting with trimethylsilylchloride. It finds application in the manufacturing of a variety of chromium-containing pigment in Paint industry and other chromium salts in the chemical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 83°C |
Color | Black to Green |
Physical Form | Crystalline |
Quantity | 100 g |
UN Number | UN3260 |
Merck Index | 14,2222 |
Solubility Information | Slightly soluble in acetone. Soluble in water,alcohol. Insoluble in ether |
IUPAC Name | trichlorochromium;hexahydrate |
Formula Weight | 266.45 (158.35 Anhydrous) |
Percent Purity | ≤99.5% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'