missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) chloride hexahydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog Number 11310939
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
10060-12-5
Cl3CrH12O6
266.44
MFCD00149660
LJAOOBNHPFKCDR-UHFFFAOYSA-K
chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
104957
CHEBI:53442
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
This item is not returnable. View return policy
10060-12-5
Cl3CrH12O6
266.44
MFCD00149660
LJAOOBNHPFKCDR-UHFFFAOYSA-K
chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
104957
CHEBI:53442
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]

 

Chromium(III) chloride hexahydrate is used as an organic synthesis catalyst and as an additive to intravenous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY
Melting Point 83°C
Color Green
Physical Form Crystalline Aggregates
Quantity 5 g
Linear Formula CrCl3·6H2O
UN Number UN3260
Merck Index 14,2222
Solubility Information Soluble in water and alcohol. Insoluble in diethyl ether.
IUPAC Name chromium(3+) hexahydrate trichloride
Formula Weight 266.45 (158.35 Anhydrous)
Percent Purity 99.998%
Grade Puratronic™
Odor Odorless
Density 1.835 g/mL
Assay (metals basis)
Chemical Name or Material Chromium(III) chloride hexahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'