missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) hexafluoro-2,4-pentanedionate, Thermo Scientific™

£84.09 - £399.00

Chemical Identifiers

CAS 14592-80-4
Molecular Formula C15H3CrF18O6
Molecular Weight (g/mol) 673.15
MDL Number MFCD00044995
InChI Key GGQIHZBUYJMHAF-UHFFFAOYSA-N
Synonym Chromium(III) hexafluoroacetylacetonate
PubChem CID 91996126
IUPAC Name chromium(3+);(Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate
SMILES [Cr+3].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11388887
View Documents
Thermo Scientific Alfa Aesar
038581.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11398887
View Documents
Thermo Scientific Alfa Aesar
038581.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

14592-80-4
673.15
GGQIHZBUYJMHAF-UHFFFAOYSA-N
91996126
[Cr+3].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F
C15H3CrF18O6
MFCD00044995
Chromium(III) hexafluoroacetylacetonate
chromium(3+);(Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate

Specifications

83°C to 88°C
Green
C15H3CrF18O6
Chromium(III) hexafluoroacetylacetonate
GGQIHZBUYJMHAF-UHFFFAOYSA-N
chromium(3+);(Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate
91996126
Odorless
14592-80-4
Crystalline
MFCD00044995
Insoluble in water
[Cr+3].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F
673.15
673.14
Chromium(III) hexafluoro-2,4-pentanedionate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications