Learn More
Chromium(III) nitrate nonahydrate, 98.5%, Thermo Scientific Chemicals
Description
Chromium nitrate is used in the manufacture of alkali metal-free catalysts and in preservation. It is used in the preparation of chrome catalysts, in textile printing operations, and as a corrosion inhibitor. Chromium(III) nitrate as a source of chromium is added to food supplements for nutritional purposes .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 60°C |
Color | Purple |
Physical Form | Crystalline |
Quantity | 25 g |
Assay Percent Range | 98.5% |
Solubility Information | Soluble in water and alcohol. |
Formula Weight | 400.14 (238.01 Anhydrous) |
Odor | Odorless |
Appearance | Purple crystalline |
Chemical Name or Material | Chromium(III) nitrate nonahydrate, nonahydrate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.