missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) nitrate nonahydrate, 98.5%

Product Code. p-3256984
Click to view available options
Quantity:
25 g
250 g
1 kg
Unit Size:
1kg
250g
25g
This item is not returnable. View return policy

Product Code. 11307297

Brand: Thermo Scientific Alfa Aesar 011565.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use As a Catalyst, Corrosion Inhibitor, in Textile Printing | CAS: 7789-02-8 | CrN3O9 | 238.01 g/mol

Chromium nitrate is used in the manufacture of alkali metal-free catalysts and in preservation. It is used in the preparation of chrome catalysts, in textile printing operations, and as a corrosion inhibitor. Chromium(III) nitrate as a source of chromium is added to food supplements for nutritional purposes .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-02-8
Molecular Formula CrN3O9
Molecular Weight (g/mol) 238.01
MDL Number MFCD00149659
InChI Key PHFQLYPOURZARY-UHFFFAOYSA-N
Synonym chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate
PubChem CID 9887206
IUPAC Name chromium(3+);trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]

Specifications

Melting Point 60°C
Color Purple
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 98.5%
Solubility Information Soluble in water and alcohol.
Formula Weight 400.14 (238.01 Anhydrous)
Odor Odorless
Appearance Purple crystalline
Chemical Name or Material Chromium(III) nitrate nonahydrate, nonahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.