missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) nitrate nonahydrate, 99%, Thermo Scientific Chemicals

Product Code. 10124990
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-02-8
MDL Number MFCD00149659
InChI Key PHFQLYPOURZARY-UHFFFAOYSA-N
Synonym chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate
PubChem CID 9887206
IUPAC Name chromium(3+);trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]

Specifications

Melting Point 60.0°C
Color Black to Purple
pH 2 to 3 (5% aq. soln.)
Physical Form Crystalline Chunks
Quantity 500 g
Assay Percent Range 98.5% min. (Iodometry)
Molecular Formula CrN3O9·9H2O
Linear Formula Cr(NO3)3·9H2O
Merck Index 14,2226
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: soluble in alcohol
Molecular Weight (g/mol) 400.14
Formula Weight 400.14
Percent Purity 99%
Packaging Plastic bottle
Chemical Name or Material Chromium(III) nitrate nonahydrate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chromium(III) nitrate nonahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'