missing translation for 'onlineSavingsMsg'
Learn More
Chromium(III) oxide, 98+%, Thermo Scientific Chemicals Product Code.: 11391629

Chromium(III) oxide, 98+%, Thermo Scientific Chemicals

Product Code. 11391629
500 g
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11391629

Brand: Thermo Scientific Alfa Aesar 012286.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is employed in metallurgical processing and has applications in chrome plating, anodizing aluminum etc… It is also used as a pigment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-38-9
Molecular Formula Cr2O3
Molecular Weight (g/mol) 151.99
MDL Number MFCD00010949
InChI Key UOUJSJZBMCDAEU-UHFFFAOYSA-N
Synonym chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxideShow MoreShow Less
PubChem CID 517277
ChEBI CHEBI:48242
SMILES [O--].[O--].[O--].[Cr+3].[Cr+3]

Specifications

Boiling Point 3000°C
Melting Point 2435°C
Color Green
Physical Form Powder
Quantity 500 g
Assay Percent Range >98%
Solubility Information Insoluble in water,alcohol,acetone and acids
IUPAC Name oxo(oxochromiooxy)chromium
Formula Weight 151.99
Odor Odorless
Refractive Index 2.551
Density 5.2 g/cm3at 20°C
Chemical Name or Material Chromium(III) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY30I055 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number012286.36,012286.A3
Lot NumberR26I021 Certificate TypeCertificate of Analysis Date09/06/2022 Catalog Number012286.36,012286.A3
Lot NumberQ25I036 Certificate TypeCertificate of Analysis Date29/04/2022 Catalog Number012286.36,012286.A3
Lot NumberQ25I026 Certificate TypeCertificate of Analysis Date29/04/2022 Catalog Number012286.36,012286.A3
Lot NumberZ29H031 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012286.36,012286.A3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromium(III) oxide, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.