missing translation for 'onlineSavingsMsg'
Learn More

Chromium(Iii) Oxide, 99.9% trace metals basis, Honeywell™

Powder, 99.9% trace metals basis

£40.53 - £139.73

Chemical Identifiers

CAS 1308-38-9
Molecular Formula Cr2O3
Molecular Weight (g/mol) 151.99
MDL Number MFCD00010949
InChI Key UOUJSJZBMCDAEU-UHFFFAOYSA-N
Synonym chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
PubChem CID 517277
ChEBI CHEBI:48242
SMILES [O--].[O--].[O--].[Cr+3].[Cr+3]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15601750
View Documents
Honeywell Research Chemicals
203068-5G
5 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15691740
View Documents
Honeywell Research Chemicals
203068-25G
25 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications

Chemical Identifiers

1308-38-9
151.99
UOUJSJZBMCDAEU-UHFFFAOYSA-N
517277
[O--].[O--].[O--].[Cr+3].[Cr+3]
Cr2O3
MFCD00010949
chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
CHEBI:48242

Specifications

4000°C
1308-38-9
Powder
MFCD00010949
chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
[O--].[O--].[O--].[Cr+3].[Cr+3]
151.99
CHEBI:48242
99.9% (trace metals basis)
Chromium(Iii) Oxide
2435°C
Green
Cr2O3
NONH for all modes of transport
UOUJSJZBMCDAEU-UHFFFAOYSA-N
oxo(oxochromiooxy)chromium
517277
151.99g/mol
5.2g/cu.cm
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-160-9

missing translation for 'rtecsNumber' : GB6475000

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.