missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) oxide, 99%, amorphous powder, Thermo Scientific Chemicals

£33.06 - £108.70

Chemical Identifiers

CAS 1308-38-9
Molecular Formula Cr2O3
Molecular Weight (g/mol) 151.99
MDL Number MFCD00010949
InChI Key UOUJSJZBMCDAEU-UHFFFAOYSA-N
Synonym chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxideShow MoreShow Less
PubChem CID 517277
ChEBI CHEBI:48242
IUPAC Name dichromium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Cr+3].[Cr+3]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10591251
SDS View Documents
Thermo Scientific Acros
192085000
500 g Glass bottle
£33.06
500g
Please to purchase this item. Need a web account? Register with us today!
11954141
SDS View Documents
Thermo Scientific Acros
192080025
2.5 kg Plastic Bottle
£108.70
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-160-9

Product Identifier
  • Chromium(III) oxide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromium(III) oxide, 99%, amorphous powder, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.