missing translation for 'onlineSavingsMsg'
Learn More
Chromium(III) oxide, 99%, Thermo Scientific Chemicals Product Code.: 11371629

Chromium(III) oxide, 99%, Thermo Scientific Chemicals

Product Code. 11371629
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11371629

Brand: Thermo Scientific Alfa Aesar 012285.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chromium(III) oxide is known as green compound which is used for buffing and polishing. It can be used as abrasives. Apart from being used as a precursor to the magnetic pigment viz., chromium dioxide, it is used in inks, glasses, and pigments (viridian). They are employed as plating agents and surface treating agents. It is a catalyst in the Phillips process of manufacture of high density polythene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-38-9
Molecular Formula Cr2O3
Molecular Weight (g/mol) 151.99
MDL Number MFCD00010949
InChI Key UOUJSJZBMCDAEU-UHFFFAOYSA-N
Synonym chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxideShow MoreShow Less
PubChem CID 517277
ChEBI CHEBI:48242
IUPAC Name dichromium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Cr+3].[Cr+3]

Specifications

Boiling Point 3000°C
Melting Point 2435°C
Color Green
Physical Form Powder
Quantity 100 g
Assay Percent Range 99%
Solubility Information Insoluble in water,alcohol,acetone and acids.
Formula Weight 151.99
Odor Odorless
Refractive Index 2.551
Appearance Pale to dark green powder
Density 5.2 g/cm3at 20°C
Chemical Name or Material Chromium(III) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP14L020 Certificate TypeCertificate of Analysis Date17/03/2025 Catalog Number012285.36,012285.22
Lot NumberM24L004 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number012285.36,012285.22
Lot NumberT26K011 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number012285.36,012285.22
Lot NumberP13K007 Certificate TypeCertificate of Analysis Date19/03/2024 Catalog Number012285.36,012285.22
Lot NumberU17I021 Certificate TypeCertificate of Analysis Date20/09/2022 Catalog Number012285.36,012285.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromium(III) oxide, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.