missing translation for 'onlineSavingsMsg'
Learn More
Chromium(III) oxide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.8% (metals basis), Thermo Scientific™ Product Code.: 15453775

Chromium(III) oxide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.8% (metals basis), Thermo Scientific™

Product Code. 15453775
1 Ea.
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy

Product Code. 15453775

Brand: Thermo Scientific Alfa Aesar 041056.KS

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Magnetron sputtering source

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 3000°C
Melting Point 2435°C
CAS 1308-38-9
Color Green
Quantity 1 Ea.
Molecular Formula Cr2O3
MDL Number MFCD00010949
Synonym chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
InChI Key UOUJSJZBMCDAEU-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[Cr+3].[Cr+3]
IUPAC Name dichromium(3+) trioxidandiide
Molecular Weight (g/mol) 151.99
PubChem CID 517277
ChEBI CHEBI:48242
Formula Weight 151.99
Percent Purity 99.8%
Odor Odorless
Density 5.22 g/mL
Assay (metals basis)
Chemical Name or Material Chromium(III) oxide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromium(III) oxide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.8% (metals basis), Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.