missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) phosphate tetrahydrate

Product Code. 11382648
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11382648

Brand: Thermo Scientific Alfa Aesar 024110.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use As a Catalyst, Pigment | CAS: 10101-59-4 | CrH8O8P | 219.026 g/mol

It is used as an additive in drugs, anti-corrosive pigments, catalysts and polymer manufacture, is also used to catalyze cation exchange in sorption reactions, as a laminated structure to dampen vibration and noise in a motor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-59-4
Molecular Formula CrH8O8P
Molecular Weight (g/mol) 219.026
MDL Number MFCD00016049
InChI Key BVZHAJJRIRDSTG-UHFFFAOYSA-K
Synonym chromium iii phosphate tetrahydrate, cr.po4.4h2o, chromium 3+ ion tetrahydrate phosphate, chromium 3+ phosphate-water 1/1/4, chromium 3+ tetrahydrate phosphate
PubChem CID 53471880
IUPAC Name chromium(3+);phosphate;tetrahydrate
SMILES O.O.O.O.[O-]P(=O)([O-])[O-].[Cr+3]

Specifications

Physical Form Powder
Quantity 50 g
Solubility Information Soluble in acids. Insoluble in water.
Formula Weight 219.04 (146.97 Anhydrous)
Appearance Dark powder
Density 2.26
Chemical Name or Material Chromium(III) phosphate tetrahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.