missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium(III) potassium sulfate dodecahydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Chromium(III) potassium sulfate is used for tanning of leather and as a photographic film hardener. It is used in manufacturing of chromium-containing catalyst.It is used in mordantin in printing and dyeing industry and it could also be used as analytical reagent. In addition, it is also used as food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chromium(III) potassium sulfate is used for tanning of leather and as a photographic film hardener. It is used in manufacturing of chromium-containing catalyst.It is used in mordantin in printing and dyeing industry and it could also be used as analytical reagent. In addition, it is also used as food additive.
Solubility
Soluble in water. 24 g/100 mL (15°C)
Notes
Incompatible with magnesium, aluminum. Store away from heat.
Chromium(III) potassium sulfate is used for tanning of leather and as a photographic film hardener. It is used in manufacturing of chromium-containing catalyst.It is used in mordantin in printing and dyeing industry and it could also be used as analytical reagent. In addition, it is also used as food additive.
Solubility
Soluble in water. 24 g/100 mL (15°C)
Notes
Incompatible with magnesium, aluminum. Store away from heat.
Specifications
Melting Point | 89°C |
Quantity | 500 g |
Linear Formula | CrK(SO4)2·12H2O |
Merck Index | 14,2229 |
Solubility Information | Soluble in water. 24g/100 mL (15°C) |
Formula Weight | 499.39 (283.21 Anhydrous) |
Percent Purity | 98% |
Odor | Odorless |
Density | 1.83 |
Chemical Name or Material | Chromium (III) potassium sulfate dodecahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'