Learn More
Chromium(III) telluride, 99.5% (metals basis), Thermo Scientific Chemicals
£63.00 - £268.00
Chemical Identifiers
CAS | 12053-39-3 |
---|---|
Molecular Formula | Cr2Te3 |
Molecular Weight (g/mol) | 486.79 |
MDL Number | MFCD00016040 |
InChI Key | PDJHBRMODJKXLQ-UHFFFAOYSA-N |
Synonym | tellanylidene tellanylidenechromio tellanyl chromium, chromium iii telluride, chromium telluride, chromium iii telluride,-325 mesh |
PubChem CID | 6381587 |
IUPAC Name | tellanylidenechromium;tellurium |
SMILES | [Te].[Cr]=[Te].[Cr]=[Te] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11309357
|
Thermo Scientific Alfa Aesar
040206.06 |
5 g |
£63.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11399347
|
Thermo Scientific Alfa Aesar
040206.14 |
25 g |
£268.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chromium telluride is used in a number of industrial applications including infrared detection and imaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12053-39-3 | |
486.79 | |
PDJHBRMODJKXLQ-UHFFFAOYSA-N | |
6381587 | |
[Te].[Cr]=[Te].[Cr]=[Te] |
Cr2Te3 | |
MFCD00016040 | |
tellanylidene tellanylidenechromio tellanyl chromium, chromium iii telluride, chromium telluride, chromium iii telluride,-325 mesh | |
tellanylidenechromium;tellurium |
Specifications
∼1300°C | |
Gray to Black | |
Cr2Te3 | |
tellanylidene tellanylidenechromio tellanyl chromium, chromium iii telluride, chromium telluride, chromium iii telluride,-325 mesh | |
PDJHBRMODJKXLQ-UHFFFAOYSA-N | |
tellanylidenechromium;tellurium | |
6381587 | |
99.5% | |
7.0 g/mL | |
Chromium(III) telluride |
12053-39-3 | |
Powder | |
MFCD00016040 | |
Insoluble in water. | |
[Te].[Cr]=[Te].[Cr]=[Te] | |
486.79 | |
486.79 | |
Odorless | |
(metals basis) |
Safety and Handling
GHS H Statement
H301-H350-H332-H317
Toxic if swallowed.
May cause cancer.
Harmful if inhaled.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P363-P501c
H302+H332-H317-H350
missing translation for 'einecsNumber' : 235-003-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only