missing translation for 'onlineSavingsMsg'
Learn More

Chromium(VI) Oxide, Certified AR for Analysis, Fisher Chemical™

£118.00 - £189.00

Chemical Identifiers

CAS 1333-82-0
Molecular Formula CrO3
Molecular Weight (g/mol) 99.99
MDL Number MFCD00010952
InChI Key WGLPBDUCMAPZCE-UHFFFAOYSA-N
IUPAC Name trioxochromium
SMILES O=[Cr](=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11463663
View Documents
Fisher Chemical
C/5920/50
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11403723
View Documents
Fisher Chemical
C/5920/53
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

1333-82-0
99.99
WGLPBDUCMAPZCE-UHFFFAOYSA-N
O=[Cr](=O)=O
CrO3
MFCD00010952
trioxochromium

Specifications

196°C
Red to Violet
Solid
MFCD00010952
O=[Cr](=O)=O
99.99
Plastic powder jar
1333-82-0
1
CrO3
WGLPBDUCMAPZCE-UHFFFAOYSA-N
trioxochromium
99.99g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Chromium(VI) oxide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 1B
  • Oxidising solid Category 1
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 A
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H350-May cause cancer.
  • H340-May cause genetic defects.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H361f-Suspected of damaging fertility.
  • H301-Toxic if swallowed.
  • H271-May cause fire or explosion; strong oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P350-IF ON SKIN: Gently wash with plenty of soap and water.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications