missing translation for 'onlineSavingsMsg'
Learn More

Chromium nitride, Thermo Scientific Chemicals

£44.12 - £112.56

Chemical Identifiers

CAS 12053-27-9
Molecular Formula CrN
Molecular Weight (g/mol) 66.00
MDL Number MFCD00016048
InChI Key YTZQKGWLQOLFNF-UHFFFAOYSA-N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11386518
View Documents
Thermo Scientific Alfa Aesar
012149.14
25 g
£44.12
25g
Please to purchase this item. Need a web account? Register with us today!
11396518
View Documents
Thermo Scientific Alfa Aesar
012149.22
100 g
£112.56
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Chromium nitride is used for stainless steel, corrosion resistance steel, alloy steel and such special steel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12053-27-9
66.00
YTZQKGWLQOLFNF-UHFFFAOYSA-N
CrN
MFCD00016048

Specifications

12053-27-9
CrN
Insoluble in water.
66.00
Powder
MFCD00016048
YTZQKGWLQOLFNF-UHFFFAOYSA-N
Chromium nitride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H317

P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c

H302-H317

missing translation for 'einecsNumber' : 235-002-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Chromium nitride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

Supplemental information
  • MIXTURE LIST-Contains : Chromium nitride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.