missing translation for 'onlineSavingsMsg'
Learn More
Chromium pieces, 3-12mm (0.12-0.47in), 99.2% (metals basis), Thermo Scientific Chemicals Product Code.: 11316577

Chromium pieces, 3-12mm (0.12-0.47in), 99.2% (metals basis), Thermo Scientific Chemicals

Product Code. 11316577
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11316577

Brand: Thermo Scientific Alfa Aesar 000472.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-47-3
Molecular Formula Cr
Molecular Weight (g/mol) 52.00
MDL Number MFCD00010944
InChI Key VYZAMTAEIAYCRO-UHFFFAOYSA-N
Synonym chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
PubChem CID 23976
ChEBI CHEBI:28073
IUPAC Name chromium
SMILES [Cr]

Specifications

Quantity 100 g
Percent Purity 99.2%
Odor Odorless
Form Pieces
Assay (metals basis)
Chemical Name or Material Chromium pieces, 3 to 12mm (0.12 to 0.47 in.)

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX23J027 Certificate TypeCertificate of Analysis Date09/11/2023 Catalog Number000472.36,000472.22
Lot NumberW20J022 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number000472.36,000472.22
Lot NumberS27J001 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number000472.36,000472.22
Lot NumberT19J018 Certificate TypeCertificate of Analysis Date25/07/2023 Catalog Number000472.36,000472.22
Lot NumberN28J039 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number000472.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromium pieces, 3-12mm (0.12-0.47in), 99.2% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.