missing translation for 'onlineSavingsMsg'
Learn More

Chromium, plasma standard solution, Specpure™ Cr 10,000 ^mg/ml, Thermo Scientific Chemicals

Product Code. 11460171
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00010944

Specifications

Concentration Cr(NO3)3 in 5% HNO3
Quantity 500 mL
UN Number UN3264
Grade Specpure™
Physical Form Liquid
Chemical Name or Material Chromium plasma standard solution, Cr 10000 μg/mL

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.