missing translation for 'onlineSavingsMsg'
Learn More

Chromium powder, -10+20 mesh, 99.996% (metals basis)

Product Code. 11385948
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11385948

Brand: Thermo Scientific Alfa Aesar 010452.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-47-3 | Cr | 52.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-47-3
Molecular Formula Cr
Molecular Weight (g/mol) 52.00
MDL Number MFCD00010944
InChI Key VYZAMTAEIAYCRO-UHFFFAOYSA-N
Synonym chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
PubChem CID 23976
ChEBI CHEBI:28073
IUPAC Name chromium
SMILES [Cr]

Specifications

Color Gray
Appearance Gray powder
Density 7.19g/cm3
Quantity 25 g
Mesh Size -10+20 Mesh
Odor Odorless
Vapor Pressure 0 hPa
Assay Percent Range 99.996% (Metals basis)
Form Powder
Chemical Name or Material Chromium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.