missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium powder, -200 mesh, 99.94% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Silver |
UN Number | UN3089 |
Quantity | 25 g |
Percent Purity | 99.95% |
Mesh Size | −200 mesh |
Odor | Odorless |
Assay | (metals basis) |
Chemical Name or Material | Chromium powder |
Safety and Handling

Product Identifier
- Chromium powder
- Warning
- Flammable solid Category 2
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- MIXTURE LIST-Contains : Chromium
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Chromium powder, -200 mesh, 99.94% (metals basis), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction