missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium selenide, Black powder, 99.5% (Metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
Description
It is used in semiconductor, chemical vapor deposition (CVD) and physical vapor deposition (PVD) display and optical applications. It is used for coating solar energy or fuel cells and flip-chip applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Black |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.5% (Metals basis) |
Solubility Information | Insoluble in water. |
Formula Weight | 130.96 |
Odor | Odorless |
Chemical Name or Material | Chromium selenide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'