Learn More
Chromium(VI) oxide, 98+%, ACS reagent, Thermo Scientific Chemicals
£97.50 - £315.00
Chemical Identifiers
CAS | 1333-82-0 |
---|---|
Molecular Formula | CrO3 |
Molecular Weight (g/mol) | 99.99 |
MDL Number | MFCD00010952 |
InChI Key | WGLPBDUCMAPZCE-UHFFFAOYSA-N |
Synonym | chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide |
PubChem CID | 14915 |
ChEBI | CHEBI:48240 |
IUPAC Name | trioxochromium |
SMILES | O=[Cr](=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10557552
|
Thermo Scientific Acros
405235000 |
500 g | Glass bottle |
£97.50
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10412975
|
Thermo Scientific Acros
405230010 |
1 kg | Glass bottle |
£151.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10771822
|
Thermo Scientific Acros
405230025 |
2.5 kg | Glass bottle |
£315.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
1333-82-0 | |
99.99 | |
WGLPBDUCMAPZCE-UHFFFAOYSA-N | |
14915 | |
trioxochromium |
CrO3 | |
MFCD00010952 | |
chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide | |
CHEBI:48240 | |
O=[Cr](=O)=O |
Specifications
196.0°C | |
Brown-Red to Purple | |
98+% | |
CrO3 | |
01,144; 02,72; 03,54; 04,96; 05,140; 07,70; 09,115; 10,99; 13,338; 14,60 | |
chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide | |
WGLPBDUCMAPZCE-UHFFFAOYSA-N | |
trioxochromium | |
14915 | |
99.99 | |
ACS Reagent | |
2.7000g/mL |
1333-82-0 | |
Solid | |
CrO3 | |
MFCD00010952 | |
15, 2239 | |
Solubility in water: 1660g/L (20°C). Other solubilities: soluble in sulfuric acid, soluble in alcohol and mineral acids | |
O=[Cr](=O)=O | |
99.99 | |
CHEBI:48240 | |
≥98% | |
2.7 | |
Chromium(VI) oxide, 98+% |
Safety and Handling
GHS H Statement:
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Suspected of damagin
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 215-607-8
missing translation for 'rtecsNumber' : GB6650000
missing translation for 'tsca' : TSCA
RUO – Research Use Only