missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium(Vi) Oxide, ReagentPlus™, Honeywell™
Click to view available options
Quantity:
5 g
100 g
1 kg
2.5 kg
Beschreibung
- ≥95% for general laboratory use.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Spezifikation
Quantity | 5 g |
UN Number | 1463 |
Formula Weight | 99.99g/mol |
Percent Purity | 99.9% (trace metals basis) |
Grade | ReagentPlus™ |
Chemical Name or Material | Chromium(Vi) Oxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'