missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Chromogenic Coliform Agar (ISO)

Product Code. p-8015226
Click to view available options
Format:
55mm Plate
90mm Plate
Fill Volume:
13 mL
17 mL
Fill Weight:
13mL
17mL
Unit Size:
180g
Pack of 10
This item is not returnable. View return policy

Product Code. 15678878

Brand: Thermo Scientific™ PO5318A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect, enumerate, and differentiate coliforms and E.coli in water samples in accordance with ISO 9308-1 with convenient easy-to-read prepared media.

Enumerate and differentiate coliforms and Escherichia coli in water samples with Thermo Scientific™ Oxoid™ Chromogenic Coliform Agar (ISO) prepared media plates, in accordance with ISO 9308-1 guidelines for water quality analysis. Target organisms are easy to differentiate, E.coli forming dark blue/violet colonies and coliforms pink to red colonies. Complete detection within 24 hours, removing the need for subculturing.

Chromogenic Coliform Agar (ISO) plates are a fast, accurate, and efficient way to detect coliforms and E.coli during microbiological quality testing of drinking water, disinfected pool water, and treatment plant samples.
  • Conforms to ISO 9308-1 guidelines
  • Easily and clearly differentiates E. coli and coliforms
  • Ready-to-use convenience of prepared media
The guideline, ISO 9308-1:2014 Enumeration of Escherichia coli and coliform bacteria –Part 1: Membrane filtration method for waters with low bacterial background flora, specifies a method based on membrane filtration, subsequent culture on a Chromogenic Coliform Agar medium, and calculation of the number of target organisms in the sample.

Chromogenic Coliform Agar (ISO) utilizes specific enzymes associated with general coliforms and E.coli for differentiation.
  • The chromogen Salmon GAL, when cleaved by D-galactosidase, produces a pink to red coloration; this coloration and enzyme are associated with coliforms.
  • E.coli contains D-galactosidase which cleaves Salmon GAL; however, it also contains D-glucuronidase, which cleaves the chromogen X-glucuronide. The combination of these chromogens produces a dark blue to violet coloration associated with E.coli.
Tergitol-7 within the medium inhibits Gram positive bacteria and Gram negative bacteria with exception of coliforms.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY

Specifications

Color Light ivory, transparent
Content And Storage 2° to 12°C, in the dark
Description Chromogenic Coliform Agar (ISO)
Format 90mm Plate
Manufacturing Origin Germany
Product Type Agar
Fill Volume 17 mL
Fill Weight 17mL
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.