missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromogranin B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14968-1-AP-150UL
This item is not returnable.
View return policy
Description
Chromogranin B is a tyrosine sulfated secretory protein found in a wide variety of peptidergic endocrine cells. Chromogranin functions as a neuroendocrine secretory granule protein which may be the precursor for other biologically active peptides.Specifications
Chromogranin B | |
Polyclonal | |
Unconjugated | |
CHGB | |
CgB, CHGB, Chromogranin B, SCG1, Secretogranin 1, Secretogranin I, SgI | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1114, 12653 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P05060, P16014 | |
CHGB | |
Chromogranin B Fusion Protein Ag6833 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Chromogranin B Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction