missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ ChromoSphere™-T Certified Size Standards
Achieve high visual contrast with Thermo Scientific™ ChromoSphere™-T Certified Size Standards, colored polymer microspheres with NIST™-traceable diameters.
£382.00 - £565.00
Specifications
Additive | May contain trace amount of dispersant |
---|---|
Certifications/Compliance | Certificate of Calibration and Traceability to NIST |
Concentration | 100% solids |
Content And Storage | Store at room temperature in a dry area. Store upright and keep bottle tightly sealed. Respiratory protection from dust is recommended. |
Contents | Dry dyed polystyrene (DVB) microspheres |
Product Code | Brand | Diameter (Metric) | Material | Color | Size Distribution | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Material | Color | Size Distribution | Price | Quantity & Availability | ||
11838223
|
Thermo Scientific™
BK050T |
50 μm | Polystyrene Divinylbenzene (PS-DVB) | Black | s = 3.6µm, 7.3% CV |
£565.00
1g |
Estimated Shipment: 04-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11858223
|
Thermo Scientific™
BK100T |
100 μm | Polystyrene Divinylbenzene (PS-DVB) | Black | s = 7.1µm, 7.5% CV |
£565.00
1g |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11878223
|
Thermo Scientific™
BK150T |
150 μm | Polystyrene Divinylbenzene (PS-DVB) | Black | s = 7.8µm, 5.2% CV |
£565.00
1g |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11898223
|
Thermo Scientific™
BK200T |
200 μm | Polystyrene Divinylbenzene (PS-DVB) | Black | s = 10µm, 5.0% CV |
£510.00
1g |
Estimated Shipment: 04-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11818233
|
Thermo Scientific™
BK300T |
300 μm | Polystyrene Divinylbenzene (PS-DVB) | Black | s = 7.5µm, 2.5% CV |
£510.00
1g |
Estimated Shipment: 04-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11838233
|
Thermo Scientific™
BK400T |
400 μm | Polystyrene Divinylbenzene (PS-DVB) | Black | s = 18µm, 4.8% CV |
£510.00
1g |
Estimated Shipment: 04-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11858233
|
Thermo Scientific™
BK500T |
500 μm | Polystyrene Divinylbenzene (PS-DVB) | Black | s = 22µm, 4.5% CV |
£510.00
1g |
Estimated Shipment: 04-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11844902
|
Thermo Scientific™
RD050T |
50 μm | Polystyrene Divinylbenzene (PS-DVB) | Red | s = 3.8µm, 7.7% CV |
£382.00
1g |
Estimated Shipment: 04-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11864902
|
Thermo Scientific™
RD100T |
100 μm | Polystyrene Divinylbenzene (PS-DVB) | Red | s = 7.5µm, 7.8% CV |
£510.00
1g |
Estimated Shipment: 04-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11884902
|
Thermo Scientific™
RD150T |
150 μm | Polystyrene Divinylbenzene (PS-DVB) | Red | s = 9.0µm, 6.0% CV |
£382.00
1g |
Estimated Shipment: 04-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11804912
|
Thermo Scientific™
RD200T |
200 μm | Polystyrene Divinylbenzene (PS-DVB) | Red | s = 9.5µm, 4.7% CV |
£510.00
1g |
Estimated Shipment: 04-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11824912
|
Thermo Scientific™
RD300T |
300 μm | Polystyrene Divinylbenzene (PS-DVB) | Red | s = 11µm, 3.5% CV |
£510.00
1g |
Estimated Shipment: 04-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11844912
|
Thermo Scientific™
RD400T |
400 μm | Polystyrene Divinylbenzene (PS-DVB) | Red | s = 18 µm, 4.4% CV |
£382.00
1g |
Estimated Shipment: 04-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11864912
|
Thermo Scientific™
RD500T |
500 μm | Polystyrene Divinylbenzene (PS-DVB) | Red | s = 24µm, 4.8% CV |
£382.00
1g |
Estimated Shipment: 04-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
Achieve high visual contrast with Thermo Scientific™ ChromoSphere™-T Certified Size Standards, colored polymer microspheres with NIST™-traceable diameters. These reference and calibration materials are used in the testing of filtration systems, vial cleaning evaluations, contamination control studies and calibration of injection devices.
- Internally dyed with deeply colored red or black dyes, resulting in very high contrast and visibility relative to most background materials
- Mean diameters traceable to the standard meter through the National Institute of Standards and Technology (NIST)
- Calibrated by optical microscopy
- Made from cross-linked polystyrene divinylbenzene (PS-DVB) copolymer
- Available as dry powders
- Easily dispersed in aqueous media with the aid of a small amount of surfactant or in lower alcohols, such as methanol or ethanol
- Minimal dye extraction when particles are suspended in pure alcohols
- Other organic solvents, such as ethers or chlorinated hydrocarbons, should be avoided because they will swell the particles and completely extract the dye.
Specifications
May contain trace amount of dispersant | |
100% solids | |
Dry dyed polystyrene (DVB) microspheres | |
Cleaning Evaluation, High Contrast Reference and Calibration | |
Size Standard Particles | |
1.05g/cm³ | |
1.59 at 589nm (25°C) |
Certificate of Calibration and Traceability to NIST | |
Store at room temperature in a dry area. Store upright and keep bottle tightly sealed. Respiratory protection from dust is recommended. | |
Designed for use as reference or calibration materials in applications where high contrast is required. | |
Polystyrene Divinylbenzene (PS-DVB) | |
Certificate of Calibration and Traceability to NIST | |
1 g | |
Product stable for a minimum of 24 months |