Learn More
Proteintech ChromoTek Actin-Chromobody™ Plasmid (TagRFP)
Actin-Chromobody™ fused to TagRFP.
Brand:  Proteintech ACR
Additional Details : Weight : 0.00240kg
Product Code. 17323283
  • £761.00 / 20µg
Estimated Shipment: 15-07-2024
to see stock.
Description
Description
  • DNA plasmid encoding for anti-Actin VHH (anti-Actin Nanobody) fused to TagRFP
Specifications
Specifications
20 μg