missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Nano-CaptureLigand™ Human Ig, Lambda-LC-specific VHH, Biotinylated
Nano-CaptureLigand for human Ig: lambda-specific single domain antibody/VHH [CTK0110], biotinylated.
£102.00 - £383.00
Specifications
Target Specificity | Lambda-LC of Human Ig lambda Antibodies |
---|---|
Conjugate | Biotin |
Clonality | Monoclonal |
Content And Storage | Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F |
For Use With (Application) | BLI, SPR, ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17353463
|
Proteintech
SHULB-1-10 |
10 μL |
£102.00
10µL |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
17363463
|
Proteintech
SHULB-1-100 |
100 μL |
£383.00
100µL |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
Description
- The Nano-CaptureLigand™ human Ig, lambda-LC captures and immobilizes human Ig lambda light chain (LC) antibodies to avidin/streptavidin coated surfaces used in BLI, SPR, and ELISA assays
- Consists of a biotinylated Nanobody that specifically binds to human Ig lambda LC antibodies with high affinity and in a site-directed manner
Specifications
Lambda-LC of Human Ig lambda Antibodies | |
Monoclonal | |
BLI, SPR, ELISA |
Biotin | |
Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F |