missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Nano-CaptureLigand™ Human IgE, Recombinant VHH, Biotinylated
Nano-CaptureLigand for human IgE: single domain antibody/VHH [CTK0111], biotinylated.
£102.00 - £383.00
Specifications
Target Specificity | Fc-fragment of Human IgG1, IgG2, IgG3, and IgG4, Fc-fragment of Rabbit IgG |
---|---|
Conjugate | Biotin |
Clonality | Monoclonal |
Content And Storage | Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F |
For Use With (Application) | BLI, SPR, ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17333463
|
Proteintech
SHUEB-1-10 |
10 μL |
£102.00
10µL |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
17343463
|
Proteintech
SHUEB-1-100 |
100 μL |
£383.00
100µL |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
Description
- The ChromoTek Nano-CaptureLigand™ human IgE, VHH, biotinylated is used for the site-directed and specific immobilization of human IgE in biosensor and ELISA assays
- It captures non-biotinylated human IgE antibodies to streptavidin/avidin
- Nano-CaptureLigand human IgE, VHH, biotinylated comprises a monoclonal biotinylated VHH/ Nanobody
Specifications
Fc-fragment of Human IgG1, IgG2, IgG3, and IgG4, Fc-fragment of Rabbit IgG | |
Monoclonal | |
BLI, SPR, ELISA |
Biotin | |
Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F |