missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Nano-CaptureLigand™ Mouse IgE, VHH, Biotinylated
Nano-CaptureLigand for mouse IgE: single domain antibody/VHH [CTK0112], biotinylated.
£102.00 - £383.00
Specifications
Target Specificity | Mouse IgE |
---|---|
Conjugate | Biotin |
Clonality | Monoclonal |
Content And Storage | Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F |
For Use With (Application) | BLI, SPR, ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17333483
|
Proteintech
SMSEB-1-10 |
10 μL |
£102.00
10µL |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
17343483
|
Proteintech
SMSEB-1-100 |
100 μL |
£383.00
100µL |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
Description
- The Nano-CaptureLigand mouse IgE for the site-directed and specific immobilization of mouse IgE antibodies to avidin/streptavidin coated surfaces used in biosensor and ELISA assays
- Consists of a biotinylated Nanobody that specifically binds to the mouse IgE subclass with high affinity and in a site-directed manner
Specifications
Mouse IgE | |
Monoclonal | |
BLI, SPR, ELISA |
Biotin | |
Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F |