missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Nano-CaptureLigand™ Mouse IgG2b, Fc-specific VHH, Biotinylated
Nano-CaptureLigand for mouse IgG2b: Fc-specific single domain antibody/VHH [CTK0106], biotinylated.
£102.00 - £383.00
Specifications
Target Specificity | Fc-fragment of Mouse IgG2b |
---|---|
Conjugate | Biotin |
Clonality | Monoclonal |
Content And Storage | Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F |
For Use With (Application) | BLI, SPR, ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17313493
|
Proteintech
SMSG2BB-1-10 |
10 μL |
£102.00
10µL |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
17323493
|
Proteintech
SMSG2BB-1-100 |
100 μL |
£383.00
100µL |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
Description
- The Nano-CaptureLigand mouse IgG2b captures and immobilizes mouse IgG2b antibodies and Fc-fragments to avidin/streptavidin coated surfaces used in BLI, SPR, and ELISA assays
- Consists of a biotinylated Nanobody that specifically binds to mouse IgG2b antibodies with high affinity and in a site-directed manner
Specifications
Fc-fragment of Mouse IgG2b | |
Monoclonal | |
BLI, SPR, ELISA |
Biotin | |
Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F |