missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chromotrope 2B, pure

Product Code. 15412132
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 300.0°C
CAS 548-80-1
Molecular Formula C16H9N3Na2O10S2
MDL Number MFCD00003951
Synonym Acid Red 176, C.I. 16575, 4,5-Dihydroxy-3-(p-nitrophenylazo)-2,7-naphthalenedisulfonic acid, disodium salt
InChI Key MQHFURHHKNKIRN-GVLVODDDSA-L
Infrared Spectrum Authentic
SMILES [Na+].[Na+].OC1=CC(=CC2=C1C(=O)\C(=N/NC1=CC=C(C=C1)[N+]([O-])=O)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O
IUPAC Name disodium (3E)-5-hydroxy-3-[2-(4-nitrophenyl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Molecular Weight (g/mol) 513.36
PubChem CID 44135472
Lambda Max 513 to 517nm (c=0.1, H2O)
Formula Weight 513.35
Loss on Drying 18% max.
Grade Pure
Quantity 5 g
Chemical Name or Material Chromotrope 2B
Show More Show Less
compliance-icons
Product Identifier
  • Chromotrope 2B
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'