missing translation for 'onlineSavingsMsg'
Learn More
Chromplete™ 2-Propanol, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™ Product Code.: 15970721

Chromplete™ 2-Propanol, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™

Product Code. 15970721 Shop All Thermo Scientific Products
4 L
Click to view available options
Quantity:
1 L
2.5 L
4 L
This item is not returnable. View return policy

Product Code. 15970721

Brand: Thermo Scientific T001044000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Isopropanol is suitable for HPLC, GC, SFC, TLC, ACS, and Spectrophotometry.

Chromplete Isopropanol meets the analytical requirements of USP and is suitable for use in:

  • HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
  • GC- FID, GC- ECD, GC-TCD
  • Pesticide Analysis by GC or HPLC
  • UV Spectrophotometry
  • Preparative chromatography
  • Supercritical Fluid and Thin Layer chromatography
  • Reactions/Analyses requiring ACS compliance
  • Extraction/Concentration Analyses
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -90°C
Absorbance ≤ 1.0, ≤ 0.20, ≤ 0.10, ≤ 0.015
CAS Min % 0.999
Color ≤ 5 APHA
Boiling Point 83°C
Quantity 4 L
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation ≤ 1.0 ppm
UN Number 1219
Water ≤0.05%
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Molecular Weight (g/mol) 60.10
Formula Weight 60.1
Percent Purity 99.9%
Grade Chromplete
Assay Minimum, GC-FID
Chemical Name or Material 2-Propanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1885655 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberT001042500,T001041000,T001044000
Lot Number1885798 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberT001042500,T001041000,T001044000
Lot Number1987384 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberT001042500,T001041000,T001044000
Lot Number2231873 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberT001042500,T001041000,T001044000
Lot Number2242778 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberT001042500,T001041000,T001044000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromplete™ 2-Propanol, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.