missing translation for 'onlineSavingsMsg'
Learn More
Chromplete™ Acetonitrile, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™ Product Code.: 15965310

Chromplete™ Acetonitrile, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™

Product Code. 15965310 Shop All Thermo Scientific Products
4 L, Aluminum Jug
Click to view available options
Quantity:
1 L
2.5 L
4 L
Packaging:
Aluminum Jug
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 15965310

Brand: Thermo Scientific T001014000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Acetonitrile is suitable for HPLC, GC, SFC, TLC, ACS, Spectrophotometry, and meets the analytical requirements of USP.

Chromplete Acetonitrile meets the analytical requirements of USP and is suitable for use in:

  • HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
  • GC- FID, GC- ECD, GC-TCD
  • Pesticide Analysis by GC or HPLC
  • UV Spectrophotometry
  • Preparative chromatography
  • Supercritical Fluid and Thin Layer chromatography
  • Reactions/Analyses requiring ACS compliance
  • Extraction/Concentration Analyses
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecnShow MoreShow Less
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -51°F
Absorbance <0.1 at 195 nm, <0.05 at 200 nm, <0.02 at 210 nm, <0.01 at 220 nm
CAS Min % 0.9995
Color ≤ 5 APHA
Density 0.775 - 0.78
Boiling Point 180°F
Quantity 4 L
Packaging Aluminum Jug
Physical Form Liquid
Refractive Index 1.3405 – 1.3425
Residue after Evaporation ≤ 0.8 ppm
Titratable Acid ≤ 0.008 mEq/g
Titratable Base ≤ 0.0006 mEq/g
UN Number 1648
Water ≤ 0.01 %
Formula Weight 41.05
Percent Purity 0.9995
Grade Chromplete
Assay Minimum, GC-FID
Chemical Name or Material Acetonitrile
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2237171 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberT001014000,T001011000,T001012500
Lot Number2411309 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberT001014000,T001011000,T001012500
Lot Number2173382 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberT001014000,T001011000,T001012500
Lot Number2359367 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberT001014000,T001011000,T001012500
Lot Number2491151 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberT001014000,T001011000,T001012500
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile Chromplete
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromplete™ Acetonitrile, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.