missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromplete™ Methanol, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™
27.00 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
4 L
Description
The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Methanol is suitable for HPLC, GC, SFC, TLC, ACS, and Spectrophotometry.
Chromplete Methanol meets the analytical requirements of USP and is suitable for use in:
- HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
- GC- FID, GC- ECD, GC-TCD
- Pesticide Analysis by GC or HPLC
- UV Spectrophotometry
- Preparative chromatography
- Supercritical Fluid and Thin Layer chromatography
- Reactions/Analyses requiring ACS compliance
- Extraction/Concentration Analyses
Specifications
Melting Point | -98°C |
Absorbance | ≤1.00, ≤ 0.23, ≤ 0.11, ≤ 0.01 |
CAS Min % | 0.999 |
Color | ≤ 5 APHA |
Density | 0.775 – 0.780 |
Boiling Point | 148.5°F |
Quantity | 2.5 L |
Packaging | Amber Glass |
Physical Form | Liquid |
Residue after Evaporation | ≤ 1.0 ppm |
Show More |
Safety and Handling
Product Identifier
- Methanol Chromplete
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'