missing translation for 'onlineSavingsMsg'
Learn More
Chromplete™ Methanol, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™ Product Code.: 15910721

Chromplete™ Methanol, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™

Product Code. 15910721 Shop All Thermo Scientific Products
4 L
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2.5 L
4 L
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15910721

Brand: Fisher Scientific Chemical Div T001024000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Methanol is suitable for HPLC, GC, SFC, TLC, ACS, and Spectrophotometry.

Chromplete Methanol meets the analytical requirements of USP and is suitable for use in:

  • HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
  • GC- FID, GC- ECD, GC-TCD
  • Pesticide Analysis by GC or HPLC
  • UV Spectrophotometry
  • Preparative chromatography
  • Supercritical Fluid and Thin Layer chromatography
  • Reactions/Analyses requiring ACS compliance
  • Extraction/Concentration Analyses
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spiritShow MoreShow Less
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Absorbance ≤1.00, ≤ 0.23, ≤ 0.11, ≤ 0.01
CAS Min % 0.999
Color ≤ 5 APHA
Density 0.775 – 0.780
Boiling Point 148.5°F
Quantity 4 L
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation ≤ 1.0 ppm
Titratable Acid ≤0.0003 meq/g
Titratable Base ≤0.0002 meq/g
UN Number 1230
Water ≤0.07%
Formula Weight 32.04
Percent Purity 99.9%
Grade Chromplete
Assay Minimum, GC-FID
Chemical Name or Material Methanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2043487 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberT001024000,T001021000,T001022500
Lot Number2039696 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberT001024000,T001021000,T001022500
Lot Number2041042 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberT001024000,T001021000,T001022500
Lot Number2400847 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberT001024000,T001021000,T001022500
Lot Number2213021 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberT001024000,T001021000,T001022500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromplete™ Methanol, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.