missing translation for 'onlineSavingsMsg'
Learn More

Chromyl chloride, 99.99% (metals basis)

Product Code. 11318508
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11318508

Brand: Thermo Scientific Alfa Aesar 040517.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 14977-61-8 | Cl2CrH4O2 | 158.926 g/mol

Highly spin-polarized chromium dioxide (CrO2) thin films were deposited on 100 TiO2 substrates by chemical vapor deposition using chromyl chloride as a precursor. Chromyl chloride (Cr02C12) reacts with cyclohexane solvent at 75 OC to give a dark precipitate along with chlorocyclohexane and a small amount of cyclohexene (in 10.0 and ca. 0.3% yields based on chromium). Chromyl chloride, CrO2C12, oxidizes cyclooctane, isobutane, and toluene under mild conditions (25-60 C). The reactions give chlorinated products (chlorocyclooctane, tert- butyl chloride, and benzyl chloride) and a dark chromium-containing precipitate. Used for Organic oxidations and chlorination, chromium coordination complexes, catalyst for polymerization of olefins. Chromyl chloride in carbon tetrachloride solution reacts in the cold with cyclohexene, cyclopentene, and 1-hexene to give the various isomeric chlorohydrins as the major products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Organic oxidations and chlorination, chromium coordination complexes, catalyst for polymerization of olefins
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14977-61-8
Molecular Formula Cl2CrH4O2
Molecular Weight (g/mol) 158.926
MDL Number MFCD00010951
InChI Key HOCPFYCTFLRIAK-UHFFFAOYSA-L
Synonym chromyl chloride
PubChem CID 101943071
IUPAC Name chromium(2+);dichloride;dihydrate
SMILES O.O.[Cl-].[Cl-].[Cr+2]

Specifications

Boiling Point 117°C
Melting Point -96.5°C
Physical Form Liquid
Quantity 2 g
UN Number UN1758
Merck Index 14,2248
Formula Weight 154.9
Percent Purity 99.99%
Odor Pungent, makes eyes water
Sensitivity Moisture Sensitive
Density 1.911g/mL
Assay (metals basis)
Chemical Name or Material Chromyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.