missing translation for 'onlineSavingsMsg'
Learn More
Chrysin, 99+%, Thermo Scientific Chemicals Product Code.: 10590181

Chrysin, 99+%, Thermo Scientific Chemicals

Product Code. 10590181
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10590181

Brand: Thermo Scientific Acros 110320050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 480-40-0
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.24
MDL Number MFCD00006834
InChI Key RTIXKCRFFJGDFG-UHFFFAOYSA-N
Synonym chrysin, 5,7-dihydroxyflavone, chrysine, 5,7-dihydroxy-2-phenyl-4h-chromen-4-one, crysin, 4h-1-benzopyran-4-one, 5,7-dihydroxy-2-phenyl, unii-3cn01f5zj5, flavone, 5,7-dihydroxy, 5,7-dihydroxy-2-phenyl-4h-1-benzopyran-4-one, chembl117Show MoreShow Less
PubChem CID 5281607
ChEBI CHEBI:75095
IUPAC Name 5,7-dihydroxy-2-phenylchromen-4-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(C=C(C=C3O2)O)O

Specifications

Melting Point 284.0°C to 290.0°C
Color Yellow
Loss on Drying 1.0% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 18,124
Merck Index 15,226
Formula Weight 254.24
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Chrysin
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0443745 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number110320250,110321000,110320050
Lot NumberA0454436 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number110320250,110321000,110320050
Lot NumberA0443745 Certificate TypeCertificate of Analysis Date01/07/2022 Catalog Number110320250,110321000,110320050
Lot NumberA0400764 Certificate TypeCertificate of Analysis Date15/03/2021 Catalog Number110320250,110321000,110320050
Product Identifier
  • Chrysin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chrysin, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.