missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHST12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15341-1-AP-150UL
This item is not returnable.
View return policy
Description
Catalyzes the transfer of sulfate to position 4 of the N-acetylgalactosamine (GalNAc) residue of chondroitin and desulfated dermatan sulfate. Chondroitin sulfate constitutes the predomit proteoglycan present in cartilage and is distributed on the surfaces of many cells and extracellular matrices. Activity toward partially desulfated dermatan sulfate is however lower. Does not form 4, 6-di-O-sulfated GalNAc when chondroitin sulfate C is used as an acceptor.Specifications
CHST12 | |
Polyclonal | |
Unconjugated | |
CHST12 | |
C4S 2, C4ST 2, C4ST2, CHST12, Sulfotransferase Hlo | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
304322, 55501, 59031 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99LL3, Q9NRB3 | |
Chst12 | |
CHST12 Fusion Protein Ag7544 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |