missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHTF18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13030-1-AP-20UL
This item is not returnable.
View return policy
Description
CHTF18, CHTF8.Specifications
CHTF18 | |
Polyclonal | |
Unconjugated | |
CHTF18 | |
C16orf41, C321D2.2, C321D2.3, C321D2.4, CHL12, CHTF18, Ctf18, hCTF18, RUVBL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
214901, 287146, 63922 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BIW9, Q8WVB6 | |
Chtf18 | |
CHTF18 Fusion Protein Ag3672 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CHTF18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction