Learn More
cIAP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24304-1-AP-150UL
 View more versions of this product
Product Code. 16832785
  • £349.00 / 150µL
Explore more special offers
Description
Description
The protein encoded by this gene is a member of a family of proteins that inhibits apoptosis by binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2, probably by interfering with activation of ICE-like proteases. The encoded protein inhibits apoptosis induced by serum deprivation but does not affect apoptosis resulting from exposure to menadione, a potent inducer of free radicals. The amino acid sequence predicts three baculovirus IAP repeat domains and a ring finger domain. Transcript variants encoding the same isoform have been identified.
Specifications
Specifications
cIAP2
Polyclonal
Unconjugated
BIRC3
AIP1, API2, Apoptosis inhibitor 2, BIRC3, C IAP2, CIAP2, HAIP1, hIAP 1, HIAP1, IAP 1, IAP homolog C, IAP1, MALT2, MIHC, RING finger protein 49, RNF49
Rabbit
Antigen Affinity Chromatography
RUO
330
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q13489
BIRC3
cIAP2 Fusion Protein Ag20143
150 μL