missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIAPIN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12638-1-AP-20UL
This item is not returnable.
View return policy
Description
CIAPIN1 is a cytokine-induced inhibitor of apoptosis with no relation to apoptosis regulatory molecules of the BCL2 or CASP families. Expression of CIAPIN1 is dependent on growth factor stimulation.Specifications
CIAPIN1 | |
Polyclonal | |
Unconjugated | |
CIAPIN1 | |
2810413N20Rik, Anamorsin, CIAPIN1, DRE2, PRO0915 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
109006, 307649, 57019 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5XID1, Q6FI81, Q8WTY4 | |
CIAPIN1 | |
CIAPIN1 Fusion Protein Ag3333 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |