Learn More
Thermo Scientific™ Cimarec™ i Benchtop Distributors (Link and Sync)
Ensure identical operating conditions for multiple stirrer installations by using these benchtop distributors, for use with Mini, Micro, Compact and Maxi series stirrers.
Brand:  Thermo Scientific™ 50091721
 View more versions of this product
Product Code. 10247081
  • £300.04 / Each
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Ensure identical operating conditions for multiple stirrer installations by using Thermo Scientific™ Cimarec™ i Benchtop Distributors (Link and Sync), for use with Mini, Micro, Compact and Maxi series stirrers.
  • Simultaneous control of 2, 4, 6, or 8 Mini, Micro or Compact series stirrers using a single Telemodul 20 C or Telemodul 40 C external controller
  • Simultaneous control of 2 or 4 Maxi Series stirrers using a single Telemodul 20 C or Telemodul 40 C external controller
  • Even-powered distribution for synchronized stirring
TRUSTED_SUSTAINABILITY
Specifications
Specifications
8-Place Distributor
Mini, Micro, Compact, and Maxi Series Stirrer
Benchtop Distributor
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.