missing translation for 'onlineSavingsMsg'
Learn More
Cinanserin hydrochloride, Tocris Bioscience™ Product Code.: 30001092

Cinanserin hydrochloride, Tocris Bioscience™

Product Code. 30001092
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 30001092

Brand: Tocris Bioscience 0460/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective 5-HT2 antagonist; also 3C-like proteinase inhibitor

5-HT2 antagonist. Also inhibits 3C-like proteinase (3CLpro; Mpro). Inhibits replication of SARS-CoV.

Chemical Identifiers

CAS 54-84-2
Molecular Formula C20H25ClN2OS
Molecular Weight (g/mol) 376.943
InChI Key LXGJPDKYMJJWRB-IERUDJENSA-N
Synonym cinanserin hydrochloride, cinnanserin, maptc, cinanserin hcl, unii-f65z32ki62, cinanserin hydrochloride usan, 2'-3-dimethylamino propyl thio cinnamanilide hydrochloride, 2'-3-dimethylamino propyl thio cinnamanilide monohydrochloride, 2-propenamide, n-2-3-dimethylamino propyl thio phenyl-3-phenyl-, monohydrochlorideShow MoreShow Less
PubChem CID 6433141
IUPAC Name (E)-N-[2-[3-(dimethylamino)propylsulfanyl]phenyl]-3-phenylprop-2-enamide;hydrochloride
SMILES CN(C)CCCSC1=CC=CC=C1NC(=O)C=CC2=CC=CC=C2.Cl

Specifications

Quantity 10 mg
Formula Weight 376.94
Percent Purity >99%
Chemical Name or Material Cinanserin hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cinanserin hydrochloride, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.