missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cinnamamide, 97%, predominantly trans, Thermo Scientific Chemicals
£41.10 - £188.00
Chemical Identifiers
CAS | 621-79-4 |
---|---|
Molecular Formula | C9H9NO |
Molecular Weight (g/mol) | 147.18 |
MDL Number | MFCD00008033 |
InChI Key | APEJMQOBVMLION-VOTSOKGWSA-N |
Synonym | cinnamamide, trans-cinnamamide, 3-phenylacrylamide, cinnamic amide, 2-propenamide, 3-phenyl, 3-phenylpropenamide, 2-benzylideneacetamide, e-cinnamamide, ginnamic acid amide, trans-cinnamoylamine |
PubChem CID | 5273472 |
ChEBI | CHEBI:76320 |
IUPAC Name | (E)-3-phenylprop-2-enamide |
SMILES | C1=CC=C(C=C1)C=CC(=O)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10615722
|
Thermo Scientific Acros
110360050 |
5 g | Glass bottle |
£41.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10158003
|
Thermo Scientific Acros
110360250 |
25 g | Glass bottle |
£188.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
621-79-4 | |
147.18 | |
APEJMQOBVMLION-VOTSOKGWSA-N | |
5273472 | |
(E)-3-phenylprop-2-enamide |
C9H9NO | |
MFCD00008033 | |
cinnamamide, trans-cinnamamide, 3-phenylacrylamide, cinnamic amide, 2-propenamide, 3-phenyl, 3-phenylpropenamide, 2-benzylideneacetamide, e-cinnamamide, ginnamic acid amide, trans-cinnamoylamine | |
CHEBI:76320 | |
C1=CC=C(C=C1)C=CC(=O)N |
Specifications
621-79-4 | |
White to Beige | |
96% min. (HPLC) | |
C6H5CH=CHCONH2 | |
09, 587 | |
APEJMQOBVMLION-VOTSOKGWSA-N | |
(E)-3-phenylprop-2-enamide | |
5273472 | |
147.18 | |
Crystalline Powder |
146.0°C to 150.0°C | |
Authentic | |
C9H9NO | |
MFCD00008033 | |
cinnamamide, trans-cinnamamide, 3-phenylacrylamide, cinnamic amide, 2-propenamide, 3-phenyl, 3-phenylpropenamide, 2-benzylideneacetamide, e-cinnamamide, ginnamic acid amide, trans-cinnamoylamine | |
C1=CC=C(C=C1)C=CC(=O)N | |
147.18 | |
CHEBI:76320 | |
97% | |
Cinnamamide, Predominantly trans |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 210-707-8
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Cinnamamide, predominantly trans
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only