missing translation for 'onlineSavingsMsg'
Learn More
Cinnamic Acid, 99+%, Technical, Fisher Chemical Product Code.: 11478123

Cinnamic Acid, 99+%, Technical, Fisher Chemical

Product Code. 11478123
100g, Plastic powder jar
Click to view available options
100g, Plastic powder jar
This item is not returnable. View return policy

Product Code. 11478123

Brand: Fisher Chemical C/6100/48

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 621-82-9
Melting Point 133°C
Color White
Packaging Plastic powder jar
Molecular Formula C9H8O2
MDL Number 4369
InChI Key WBYWAXJHAXSJNI-VOTSOKGWSA-N
Molecular Weight (g/mol) 148.16
Formula Weight 148.0548g/mol
Physical Form Solid
compliance-icons
Product Identifier
  • CINNAMIC ACID
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cinnamic Acid, 99+%, Technical, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.