missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl alcohol, 98%

Product Code. 11489943
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11489943

Brand: Thermo Scientific Alfa Aesar A13025.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-54-1 | C9H10O | 134.178 g/mol

Cinnamyl alcohol is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in perfumery and as well as a deodorant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cinnamyl alcohol is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in perfumery and as well as a deodorant.

Solubility
Soluble in water 1.8 g/L (20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-54-1
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00002921
InChI Key OOCCDEMITAIZTP-QPJJXVBHSA-N
Synonym cinnamyl alcohol, cinnamic alcohol, 3-phenyl-2-propen-1-ol, 3-phenylprop-2-en-1-ol, zimtalcohol, styryl carbinol, e-3-phenylprop-2-en-1-ol, 3-phenylallyl alcohol, trans-cinnamyl alcohol, styryl alcohol
PubChem CID 5315892
ChEBI CHEBI:33227
IUPAC Name (E)-3-phenylprop-2-en-1-ol
SMILES C1=CC=C(C=C1)C=CCO

Specifications

Melting Point 32°C to 35°C
Density 1.044
Boiling Point 257°C to 258°C
Flash Point 126°C (258°F)
Odor Pleasant
Linear Formula C6H5CH=CHCH2OH
Quantity 1000 g
Beilstein 1903999
Merck Index 14,2302
Solubility Information Soluble in water 1.8g/L (20°C).
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material Cinnamyl alcohol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.